Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldolase B Mouse anti-Human, DyLight 488, Clone: OTI3C6, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270162G
Description
Aldolase B Monoclonal antibody specifically detects Aldolase B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Aldolase B | |
OTI3C6 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Affinity Purified, Conjugated | |
ALDOB | |
Full length recombinant protein of human Aldolase B (NP_000026) produced in HEK293 cell. | |
Immunogen affinity purified | |
Cancer | |
Primary | |
229.0 |
Flow Cytometry, Immunohistochemistry, Western Blot | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ALDO2, aldolase B, fructose-bisphosphate, EC 4.1.2.13, fructose-bisphosphatase, fructose-bisphosphate aldolase B | |
Mouse | |
IgG2b | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title