Learn More
Aldolase C Mouse anti-Rat, Alexa Fluor™ 594, Clone: 1A1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP225144AF594
Catalog No. NP22144A94
Add to cart
Description
Description
Aldolase C Monoclonal antibody specifically detects Aldolase C in Rat samples. It is validated for Western Blot,Flow Cytometry,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
Aldolase C
1A1
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
ALDC, aldolase 3, aldolase C, fructose-bisphosphate, Brain-type aldolase, EC 4.1.2.13, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Rat
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
ALDOC
The C-terminal 23 amino acids of human Aldolase C (KYEGSGEDGGAAAQSLYIANHAY). [UniProtNo. P09972]
0.1 mL
Primary
230