Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldon™ Innovating Science™ Oil Spill Cleanup: Biological vs. Physical Kit
SCI_ED

Ideal for students to compare biological and physical methods of oil spill cleanup.

Supplier:  Aldon™ IS9705

Catalog No. S06969


Add to Cart

Alert:

WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, which is known to the State of California to cause cancer.


Includes:
  • Oil degrading microbe blend (15g)
  • Oil degrading microbe blend
  • Nutrient QuickSolution, to make 1000mL (one)
  • Hydrocarbon encapsulating powder (200g)
  • Plastic vials (32)
  • Plastic cups
  • Cotton balls (one bag)
  • Teacher's Manual and Student Study Guide copymasters

Description

Description

  • Examine effectiveness of each approach using oil degrading microbes blend and hydrocarbon encapsulating polymer to oil
  • Specially prepared stained vegetable oil to simulate crude oil
  • Eliminate associated disposal costs of the real thing which still allowing students to perform procedure using real oil

Order Info

For 15 groups

Specifications

Specifications

Oil Spill Cleanup Kit
7 to 14
For students to compare biological and physical methods of oil spill cleanup
1
White
13 to 20
15g oil degrading microbe blend, 250ml simulated crude oil, 1 nutrient quicksolution, to make 1000ml, 200g hydrocarbon encapsulating powder, 32 plastic vials, 15 plastic cups, 1 bag cotton balls
15 Lab Groups
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit