Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldosterone Antibody (BGN/10/42133), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062317JF549
Description
Aldosterone Monoclonal antibody specifically detects Aldosterone in All samples. It is validated for ELISASpecifications
Aldosterone | |
Monoclonal | |
Janelia Fluor 549 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
ELISA | |
BGN/10/42133 | |
50mM Sodium Borate | |
Aldosterone conjugated to Bovine Serum Albumin. | |
0.1 mL | |
Lipid and Metabolism | |
All species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction