Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldosterone Antibody, DyLight 650, Novus Biologicals™
Sheep Polyclonal Antibody
Supplier: Novus Biologicals NB10062320C
Description
Aldosterone Polyclonal antibody specifically detects Aldosterone in All samples. It is validated for ELISASpecifications
Aldosterone | |
Polyclonal | |
50mM Sodium Borate | |
Made to Broad ALDOSTERONE | |
0.1 mL | |
Lipid and Metabolism | |
All species | |
Purified |
ELISA | |
DyLight 650 | |
Sheep | |
Immunogen affinity purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction