Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldosterone Sheep anti-Human, Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody has been used in 1 publication
Specifications
Antigen | Aldosterone |
---|---|
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10062321
|
Novus Biologicals
NB10062321 |
0.5 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Aldosterone Polyclonal specifically detects Aldosterone in Human, Rat samples. It is validated for ELISA.Specifications
Aldosterone | |
ELISA | |
Unconjugated | |
RUO | |
Human | |
Made to Human ALDOSTERONE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
ELISA 1:100-1:2000 | |
Polyclonal | |
Sheep | |
Lipid and Metabolism | |
Whole antisera with 0.09% Sodium Azide | |
IgG | |
Unpurified | |
NB100-62321 is specific for Aldosterone. Aldosterone is a steroid hormone which is produced by the adrenal gland. It has a number of functions relating to the kidney nephron including the balance of sodium and potassium, and increasing blood pressure. It is involved in two disorders: Addison's disease, where it is reduced, and Conn Syndrome, were it is it found in increased amounts. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title