Learn More
MilliporeSigma™ Supelco™ Aldrich™ Flask-Type Sprayer
Connect using the barb end to a low-pressure gas or air source of <5 psi.
Manufacturer:  MilliporeSigma™ Supelco™ Z1903731EA
 View more versions of this product
Catalog No. 11-101-0194
Add to cart
Description
Description
By positioning your thumb over the vent hole, you can control the spray pattern.
Specifications
Specifications
75 mL
TLC
Aldrich
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC