Learn More
MilliporeSigma™ Supelco™ Aldrich™ Flask-Type Sprayer
Connect using the barb end to a low-pressure gas or air source of <5 psi.
$157.90 - $176.40
Specifications
For Use With (Application) TLC
Product Line Aldrich
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
11-101-0194
View Documents
MilliporeSigma™ Supelco™
Z1903731EA
75 mL
Each for $157.90
Add to cart
11-101-0043
View Documents
MilliporeSigma™ Supelco™
Z1291781EA
250 mL
Each for $176.40
Add to cart
Description
Description
By positioning your thumb over the vent hole, you can control the spray pattern.
Specifications
Specifications
TLC
Aldrich
SDS
Product Certifications
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC