Learn More
Research Products International CorpSupplier Diversity Partner AlloGel Magenta Powder-Free Nitrile Exam Gloves, Large, 100 Gloves per Box, 10 Boxes per Case DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner 036800LCS
AlloGel coating and powder free, AlloGena magenta color gloves, offers you all the ultra comfort with protection from punctures and chemical or bloodborne pathogens without latex allergy concerns. AlloGel naturally is your answer to perfect hands by caring for them with the following ingredients: Allantoin, Collagen and Silicone. Ultra Soft-Stretch provides excellent comfort and donning, along with superior fit. Unique formulation reduces hand fatigue. Micro-textured finish ensures slip-resistance, wet or dry. Superior tactile sensitivity for medical use, such as handling delicate instruments.
Catalog No. 50-476-423