Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Afinion HbA1c Control

For use with the Afinion™ AS100 Analyzer and the Afinion™ HbA1c test

Manufacturer:  Abbott™ 1115178

Catalog No. 23-046-643

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Includes: Kit contents
  • 1 Control C I: Stabilized preparation from porcine whole blood (1 x 0.5 mL).
  • 1 Control C II: Stabilized preparation from human whole blood (1 x 0.5 mL).
  • 1 Package Insert
This item is not returnable. View return policy

Description

Description

  • Contains liquid preparations of stabilized porcine whole blood (Control C I) and human whole blood (Control C II)
  • Designed for use with the Afinion™ AS100 Analyzer and the Afinion™ HbA1c test
  • Quality control testing should be done to confirm that your Afinion™ Analyzer System is working properly and provides reliable results
  • Only when controls are used routinely and the values obtained are within acceptable ranges can accurate results for patient samples be assured

Specifications

Specifications

Healthcare
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit