Learn More
Alere Afinion™ HbA1c Test Cartridge
Gives you reliable results conveniently available when and where you need them. Patient consultations can be carried out with confidence. The HbA1c Test Cartridge contains all reagents necessary for the measurement of glycated hemoglobin.
Supplier: Alere 1115015
Description
Gives you reliable results conveniently available when and where you need them. Patient consultations can be carried out with confidence. The HbA1c Test Cartridge contains all reagents necessary for the measurement of glycated hemoglobin.
- Used to monitor metabolic control in patients with diabetes
- Aids in the diagnosis of diabetes and in identifying patients who may be at risk for developing diabetes
- Sample volume: 1.5 μL
- Assay time: 3 minutes
- Measuring range: 4 - 15% HbA1c, 20-140 mmol/mol
- Hb-range: 6 - 20 g/dL
- Total imprecision, external studies: < 3% CV
- Traceable to the IFCC Reference Method
- Reports DCCT / UKPDS aligned values
- No influence from Hb variants like HbC, HbD, HbE, HbF, HbJ and HbS
- No influence from carbamylated or pre-glycated hemoglobin
- Kit size: 15 tests. Separately packed cartridges
- Afinion™ HbA1c Control: 2 levels, ready to use
- Sample material: Capillary blood or anticoagulated venous blood (EDTA, citrate or heparin)
- Total imprecision: <3% CV
- Store test kit refrigerated (2° to 8°C) until the expiration date or at room temperature for a maximum of 90 days
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.