Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abbott Clearview H. pylori Test
Innovative test uses rapid, two-step technology for the direct detection of Helicobacter pylori antibodies
Manufacturer: Abbott 4581505020
Description
- CLIA Complexity: Waived for whole blood, Non-Waived for serum, plasma
- Sensitivity: 89%
- Specificity: 89%
- Results in 10 minutes
Specifications
H. pylori Antibodies | |
Built in control | |
H. pylori Test Kit | |
30 Cassettes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title