Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abbott™ Flu Starter Kit
Flu Starter Kit combines the DIGIVAL™ Complete Kit (Cat No 23046680) with BinaxNOW™ Influenza A and B Card 2 Test Cards (23046682) and an optional thermal printer (23046687) for all-in-one influenza testing.
Specifications
Content And Storage | Room Temperature |
---|---|
CE Marker | No |
Clia Complexity | Waived |
Detectable Analytes | Influenza A and B |
Format | Card |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
23-046-694
|
Abbott™
ARFLUBDPRT |
DIGIVAL Complete Kit; 12 A and B Test Card Sets; Thermal Printer |
Pack of 264 for $5,821.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-046-693
|
Abbott™
ARFLUBD |
DIGIVAL Complete Kit; 12 A and B Test Card Sets |
Pack of 264 for $5,375.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-046-692
|
Abbott™
ARFLUPRT |
DIGIVAL Complete Kit; 3 A and B Test Card Sets; Thermal Printer |
Pack of 66 for $4,089.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-046-691
|
Abbott™
ARFLUSTART |
DIGIVAL Complete Kit; 3 A and B Test Card Sets |
Pack of 66 for $3,583.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Immunochromatographic assay for the qualitative detection of influenza A and B nucleoprotein antigens in nasopharyngeal swabs and nasal swabs.Order Info
Order eligiblity is based upon executed contractual agreement. Please reach out to your Fisher Healthcare Account Representative for more information.
Specifications
Room Temperature | |
Waived | |
Card | |
Lateral Flow Assay | |
Influenza testing | |
22 Cards/Set |
No | |
Influenza A and B | |
Nasal and NP swabs; Nasal wash/aspirate samples | |
LF test card | |
Box | |
15 min. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title