Alere HemoPoint™ H2 Photometer Accessories

Accessories for use with Alere HemoPoint H2 Photometer (Cat. No. 23-763-162).

Manufacturer: Alere 88772

 View more versions of this product

Catalog No. 23-763-165

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Certifications/Compliance CLIA waived/professional use only, Biohazard Class II
    For Use With HemoPoint™ H2 photometer
    Includes Set of two dropper bottles: 1 x 1.5mL-low, 1 x 1.5mL-high
    Storage Temperature (Metric) 2° to 8°C
    Type Hemoglobin controls
    Capacity (Metric) 1.5mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit