Alere HemoPoint™ H2 Photometer and Starter Kits

Accurate hemoglobin and hematocrit results with one simple test

Manufacturer: Alere 55118SK400

 View more versions of this product

Catalog No. 23-763-171

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Calibration Yes
    Sample Type Capillary, venous or arterial Blood
    Certifications/Compliance CLIA waived/professional use only
    Includes HemoPoint H2 meter, 400 HemoPoint° H2 microcuvettes
    Sample Size 8μL
    Type HemoPoint H2 starter kit II
    For Use With (Application) Accurate hemoglobin and hematocrit results with one simple test

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit