Learn More
Abbott™ ID NOW Influenza A and B Control Swab Kit
Provides molecular results in less than 15 minutes, allowing you to make effective clinical decisions sooner. Abbott ID NOW is a rapid, instrument-based system for the qualitative detection of infectious diseases using isothermal nucleic acid amplification technology.
Manufacturer:  Abbott™ 425024
 View more versions of this product
Catalog No. 23-046-618
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. View substitute products »
This item is not returnable. View return policy
Description
Description
Transforming near-patient testing
  • Unique isothermal amplification technology giving you molecular results, faster than ever before
Improving patient outcomes
  • Giving you the confidence to make effective clinical decisions sooner
Optimizing operational efficiency
  • Enables prompt initiation of infection control measures
  • Connected results direct to your network
  • Facilitating effective patient management
Enabling effective Antimicrobial Stewardship
  • Molecular sensitivity in an actionable timeframe allows the prudent use of antibiotics and antivirals
A positive impact on your facility
  • Reliable near-patient testing reduces overall healthcare costs
Confidence at the point of care
  • Intuitive and reliable, allowing you to respond quickly and accurately in any setting
  • Simple and easy to use molecular diagnostic test
  • Sample type nasal swab
  • Requires only minimal training
  • Small footprint can be used in any healthcare setting
  • Streamlined workflow connectivity can be provided to your network
  • Demonstrates impressive sensitivity for the detection of both influenza A and B in clinical respiratory specimens
Specifications
Specifications
Influenza testing
Abbott ID NOW Influenza A and B Test Kit