Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ ID NOW Influenza A and B Control Swab Kit Available on GSA/VA Contract for Federal Government customers only.

Provides molecular results in less than 15 minutes, allowing you to make effective clinical decisions sooner. Abbott ID NOW is a rapid, instrument-based system for the qualitative detection of infectious diseases using isothermal nucleic acid amplification technology.

Specifications

For Use With (Application) Influenza testing
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
23-046-619 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Abbott™
425080
Abbott ID NOW Influenza A and B Control Swab Kit
Pack of 20 for $447.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
23-046-618
View Documents
Abbott™
425024
Abbott ID NOW Influenza A and B Test Kit This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. View substitute products »
Description

Description

Transforming near-patient testing

  • Unique isothermal amplification technology giving you molecular results, faster than ever before
Improving patient outcomes
  • Giving you the confidence to make effective clinical decisions sooner
Optimizing operational efficiency
  • Enables prompt initiation of infection control measures
  • Connected results direct to your network
  • Facilitating effective patient management
Enabling effective Antimicrobial Stewardship
  • Molecular sensitivity in an actionable timeframe allows the prudent use of antibiotics and antivirals
A positive impact on your facility
  • Reliable near-patient testing reduces overall healthcare costs
Confidence at the point of care
  • Intuitive and reliable, allowing you to respond quickly and accurately in any setting
  • Simple and easy to use molecular diagnostic test
  • Sample type nasal swab
  • Requires only minimal training
  • Small footprint can be used in any healthcare setting
  • Streamlined workflow connectivity can be provided to your network
  • Demonstrates impressive sensitivity for the detection of both influenza A and B in clinical respiratory specimens

 

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Influenza testing
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit