Alere iCassette™ Single Test Drug Screen Cassettes
Rapid and accurate screening of drugs in minutes
Manufacturer: Alere IDAM102
 View more versions of this product
Catalog No. 22-911-099
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For screening single drug in urine sample
    Quantity 25/Pk.
    Product Type Amphetamine (AMP) Single Test Cassette,Amphetamine (AMP) Single Test Cassette
    • Available as single drug testing cassette
    • Requires small amount of urine (3mL) to run
    • Pipet included for procedure testing
    • Administrator remains in control of test device
  • Description & Specifications
Related Products
Product Content Feedback