Learn More
Alere iCup? Dx Pro Urine Drug Screening Device
Easy to use, non-tilt auto activated cup for screening a wide range of drugs
Manufacturer:  Alere IDXP112702
 View more versions of this product
Catalog No. 22-911-066
  Check Availability
Add to cart
Specifications
Description
  • Self-contained cup is ideal for sending presumptive positive specimens to lab for confirmation
  • Low-level opiate cutoffs (300ng/mL)
  • Available with specimen validity test (SVT) and temperature strip help to monitor specimen tampering (SVT includes SG, OX, pH)
  • Simple to use device gives results in five minutes
  • CLIA waived
Description & Specifications
Specifications
AMP, BAR, BZO, COC, mAMP, MDMA, MTD, MOP(OPI300), OXY, PCP, TCA, THC