Alere iCassette™ Urine Drug Screen
One step screening test for the simultaneous detection of multiple drugs and metabolites in urine
Manufacturer: ALERE INSTANT TECHNOLOGIES INC IDOA1165
 View more versions of this product
Catalog No. 22-911-022
  Check Availability
Add to Cart
    Specifications
    Product Type iCassette™ Urine drug screen
    Detectable Analytes COC, THC, OPI, AMP, mAMP, PCP
    Quantity 25 Pack
    Sample Type Urine
    Test Time 5 min.
  • Description & Specifications
Related Products
Product Content Feedback