Learn More
Abbott™ iCup™ Dx CLIA-Waived Tilt and Read Urine Drug Screens
Tests for up to 12 drugs at once
Manufacturer:  Alere IDXA1127023
 View more versions of this product
Catalog No. 22-911-016
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Detect the presence of drugs and adulterants in just minutes
  • The iCup Dx Drug Screen offers versatility with up to 12 drug assays in one cup
  • FDA 510(k) Cleared and CLIA Waived
  • Multiple configurations available
  • Built-in validity screening to alleviate tampering concerns
  • Room temperature storage
  • Comes with temperature strip to verify range of collected sample
  • Leakproof, secure collection cup; just tilt after collection and read at 5 minutes
  • Ideal for sending presumptive positive specimens to the lab for confirmation
Specifications
Specifications
Urine
COC, THC, OPI, AMP, mAMP, PCP, BZO, BAR, OXY, MTD, TCA, MDMA (Bl, Cr, Ni, pH, SG)
iCup™ Dx Urine drug screen panel
25 Pack