Learn More
Alere iCup™ Dx CLIA-Waived Tilt and Read Urine Drug Screens
Tests for up to 12 drugs at once
Manufacturer:  ALERE INSTANT TECHNOLOGIES INC IDXA187016
 View more versions of this product
Catalog No. 22-911-014
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Waived
25 Pack
5 min.
COC, THC, OPI, AMP, mAMP, BZO, OXY, MTD (Bl, Cr, Ni, pH, SG)
Urine
iCup™ Dx Urine drug screen panel