Alere iCup™ Dx CLIA-Waived Tilt and Read Urine Drug Screens
Tests for up to 12 drugs at once
Manufacturer: ALERE INSTANT TECHNOLOGIES INC IDXA1107142
 View more versions of this product
Catalog No. 22-911-015
  Check Availability
Add to Cart
    Specifications
    Product Type iCup™ Dx Urine drug screen panel
    Detectable Analytes COC, THC, OPI, AMP, mAMP, MDMA, BAR, BZO, MTD, OXY (Cr, Ni, pH, Bl, SG)
    Quantity 25 Pack
    Sample Type Urine
    Test Time 5 min.
    Clia Complexity Waived
  • Description & Specifications
Related Products
Product Content Feedback