Alere iScreen™ Dip Drug Cards
Testing for up to 12 drugs on one device
Manufacturer: ALERE INSTANT TECHNOLOGIES INC IS5MB
 View more versions of this product
Catalog No. 22-911-034
  Check Availability
Add to Cart
    Specifications
    Product Type iScreen Urine drug screen
    Detectable Analytes COC, THC, MOP, mAMP, BZO
    Quantity 25/Pack
    Sample Type Urine
    Test Time Results in minutes
  • Description & Specifications
Related Products
Product Content Feedback