Alere iScreen™ Dx Dip Cards
Urine drug screening device with multi-drug configurations to meet clinical testing needs
Manufacturer: Alere IS10DX
 View more versions of this product
Catalog No. 22-911-060
  Check Availability
Add to Cart
Disclaimers:
For in-vitro diagnostic use only
    Specifications
    Detectable Analytes AMP,BAR,BZO,COC,mAMP,MOP,MTD,OXY,TCA,THC
    Product Type Multi-drug configuration
  • Description & Specifications
Related Products
Product Content Feedback