Learn More
Alere iScreen? Dx Dip Cards, Single Drug Configuration
For rapid urine drug screening (single)
Manufacturer:  Alere IS1 MOPDX
 View more versions of this product
Catalog No. 22-911-086
  Check Availability
Add to cart
Specifications
Description
  • CLIA-waived device for screening of drugs in urine sample
  • Simple test gives results as little as five minutes
  • Test device remains in control of administrator
Description & Specifications
Specifications
For urine drug screening
25 Tests/Box
Morphine (MOP-300)