Alere iScreen™ Dx Dip Cards, Single Drug Configuration
For rapid urine drug screening (single)
Manufacturer: Alere IS1 OXYDX
 View more versions of this product
Catalog No. 22-911-089
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For urine drug screening
    Quantity 25 Tests/Box
    Product Type Oxycodone (OXY),Oxycodone (OXY)
    • CLIA-waived device for screening of drugs in urine sample
    • Simple test gives results as little as five minutes
    • Test device remains in control of administrator
  • Description & Specifications
Related Products
Product Content Feedback