Alere Autoimmune ELISA Tests
An extensive line of autoimmune ELISA test kits from Wampole Laboratories. Uses substrate, stop solution, and wash buffer that are interchangeable with other Wampole assays, allowing ease-of-automation with microtiter processing systems.
Manufacturer: Alere 3503US
 View all options for this product
Catalog No. 23-047-200
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to for assistance. 
    Includes 96-well microplate (12 strips of 8 break-apart wells), SAVe Diluent™ liquid reagents, conjugate, positive and negative control, TMB substrate, stop solution, Wash Buffer
    Quantity 96 Tests
    Product Type tTg - IgA
    • Choice of incubation protocols—manual or automated—all in one kit
    • One of the broadest menus in the industry
    • Breakaway, removable test wells in a microplate format; color-coded ready-to-use reagents are provided
    • SAVe Diluent™ in most ELISA II IgG tests, to provide visual sample verification
    • Test kits are capable of being run in a manual, semi-automated, or fully automated capacity with customized instrumentation packages available to suit your laboratory's needs
  • Description & Specifications
Related Products
Product Content Feedback