Alere Autoimmune ELISA Tests Available on GSA/VA Contract for Federal Government customers only.
An extensive line of autoimmune ELISA test kits from Wampole Laboratories. Uses substrate, stop solution, and wash buffer that are interchangeable with other Wampole assays, allowing ease-of-automation with microtiter processing systems.
Manufacturer: Alere 438370
 View more versions of this product
Catalog No. 23-046-805
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Product Type ENA/Cytoplasmic -Mitochondria
    Detectable Analytes Mitochondria
    Includes 96-well microplate (12 strips of 8 break-apart wells), SAVe Diluent™ liquid reagents, conjugate, positive and negative control, TMB substrate, stop solution, Wash Buffer
    Quantity 96 Tests
  • Description & Specifications
Related Products
Product Content Feedback