Learn More
Alere Autoimmune ELISA Tests Available on GSA/VA Contract for Federal Government customers only.
An extensive line of autoimmune ELISA test kits from Wampole Laboratories. Uses substrate, stop solution, and wash buffer that are interchangeable with other Wampole assays, allowing ease-of-automation with microtiter processing systems.
Manufacturer:  Alere 2Z9681G
 View more versions of this product
Catalog No. 23-046-843
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
96-well microplate (12 strips of 8 break-apart wells), SAVe Diluent™ liquid reagents, conjugate, positive and negative control, TMB substrate, stop solution, Wash Buffer
96 Tests
ANCA Screen II Test System