Learn More
C. DIFF CHEK™ - 60 Available on GSA/VA Contract for Federal Government customers only.
C. DIFF CHEK™ - 60 is an ELISA for detecting glutamate dehydrogenase (GDH), also known as the C. difficile antigen, in stool specimens. Developed and Manufactured by TECHLAB, Inc.
Manufacturer:  Abbott Rapid Diagnostics 30392
Catalog No. WA30392
Add to cart
Alert:
The products indicated contain mercury. Do not put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by local, state or federal laws. Some products containing mercury are not available for sale in some states.
Includes: 12 x 8-well strips with breakaway wells, sample diluent, controls and calibrators, conjugate, TMB substrate and wash buffer
This item is not returnable. View return policy
Description
Description
  • Assay results in approximately 30 minutes following rapid protocol.
  • No gray zone eliminates equivocal results
  • Visual and instrument read options
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Qualitative detection of GDH antigen.
12 x 8-well strips with breakaway wells, sample diluent, control, calibrators, conjugate, TMB substrate, Wash Buffer
Clostridium difficile CHEK-60
96 Tests