Learn More
Abbott™ ColorCard™ RF Rapid Latex Test Kit
Consists of sheep erythrocytes sensitized with rabbit gamma globulin
Manufacturer:  Abbott™ 36045
Catalog No. WA-36045
Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Reagent, positive and negative controls, pipets/stirrers, disposable cards, and instructions for use
This item is not returnable. View return policy
Description
Description
  • CDC Analyte Identifier Code: 5508
  • Rapid, color-enhanced hemagglutination test for the qualitative and semiquantitative detection of rheumatoid factor (RF) in human serum
  • Read results in two minutes
  • Positive results: agglutinated reagent becomes visibly blue against a yellow background; negative results: no agglutination or color change
  • Semiquantitative results: when positive samples are examined by serial dilution, the titer is the reciprocal of the last dilution which produces a positive result (agglutination)
  • For in vitro diagnostic use
Specifications
Specifications
Non-waived
RF Rapid Latex Test Kit
50 Tests
RF in Human Serum
Reagent, positive and negative control, pipets/stirrers, disposable cards and instructions for use
2 min.