Learn More
Alere Epstein-Barr Virus (EBV) ELISA Test Kits Available on GSA/VA Contract for Federal Government customers only.
Provide increased reliability and run-to-run consistency. Use substrate, stop solution, and wash buffer that are interchangeable with other Wampole assays. This uniformity of reagents allows for ease-of-automation with microtiter processing systems.
Manufacturer:  Alere 9Z9201G
 View more versions of this product
Catalog No. 23-046-813
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Description
  • Choice of incubation protocols—manual or automated—all in one kit
  • Breakaway, removable test wells in a microplate format
  • Color-coded, ready-to-use reagents are provided
  • SAVe Diluent™ provides visual sample verification
  • Human sera
  • For in vitro diagnostic use
Description & Specifications
Specifications
Non-waived
Use substrate, stop solution and Wash Buffer that are interchangeable with other Wampole™ assays
Epstein-Barr Virus VCA IgG II ELISA Test Kit
EBV-VCA IgG II
96 Tests