Learn More
FECAL-QUIK-PREP™ Available on GSA/VA Contract for Federal Government customers only.
Fecal-Quik-Prep is a no-measuring dilution and dispensing device for stool preparation. For use with C. DIFF CHEK™ - 60 and C. DIFF TOX A/B II™. Developed and Manufactured by TECHLAB, Inc.
Manufacturer:  Abbott Rapid Diagnostics 30398
Catalog No. WA30398
Add to cart
Alert:
The products indicated contain mercury. Do not put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by local, state or federal laws. Some products containing mercury are not available for sale in some states.
Includes: 12 x 8-well strips with breakaway wells, sample diluent, controls and calibrators, conjugate, TMB substrate and wash buffer.
This item is not returnable. View return policy
Description
Description
  • The Fecal-Quik-Prep device allows for minimal contact with stool specimen.
  • Stool sample and swap are enclosed in the transport tube.
  • Premeasured diluent reduces the number of steps for testing.
  • Diluted specimens can be stored in the device for up to 72 hours.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Healthcare
C. Diff CHEK-60 30392/B and C. Diff TOX A/B II 30397/B
Fecal-Quik Prep Kit
Qualitative detection of C. difficile Antigens
96 Fecal Quik Prep devices
96 Tests