Learn More
Alere Measles (Rubeola) ELISA Test Kits Available on GSA/VA Contract for Federal Government customers only.
Provide increased reliability and run-to-run consistency. Use substrate, stop solution, and wash buffer that are interchangeable with other Wampole assays. This uniformity of reagents allows for ease-of-automation with microtiter processing systems.
Manufacturer:  Alere 9Z9271GB
 View more versions of this product
Catalog No. 23-046-826
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Description
  • Choice of incubation protocols–manual or automated–all in one kit
  • Breakaway, removable test wells in a microplate format
  • Color-coded, ready-to-use reagents are provided
  • SAVe Diluent™ provides visual sample verification
  • Individual serum specimens may be used for determination of immune status
  • CPT Code: 86765
  • For in vitro diagnostic use
Description & Specifications
Specifications
Non-waived
Scl-70 Antibodies and individual Serum specimens may be used for determination of immune status
Well strips with breakaway wells, control and calibrators, TMB substrate, stop solution, directional insert, sample SAVe Diluent™, conjugate and Wash Buffer
IgG Wampole™ Measles (Rubeola) ELISA Test Kit
86765
Microplate
480 Tests