Learn More
Jackson Immuno Research Labs Alexa Fluor 488 AffiniPure F(ab')₂ Fragment Goat Anti-Rabbit IgG, Fc fragment specific, .75mg
SDP
Supplier:  Jackson Immuno Research Labs 111546046
Encompass_Preferred
Alexa Fluor 488-AffiniPure F(ab')2 Fragment Goat Anti-Rabbit IgG, Fc Fragment Specific (min X Hu Sr Prot)
Catalog No. NC0471807