Learn More
Alfa Aesar™ Cleaning Kit for Laboratory Crucibles
For safe cleaning of crucibles without damage
Manufacturer:  Alfa Aesar™ 37995KT
Catalog No. AA37995KT
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
  • Reduces sample contamination
  • Includes liquid cleaner, variety of abrasive materials and instructions
  • For cleaning most lab crucibles, including metallic crucibles such as zirconium, nickel, molybdenum, tantalum and platinum, and crucibles of ceramic or glass
Specifications
Specifications
Liquid cleaner, variety of abrasive materials and instructions
1/ea.
Safety and Handling
Safety and Handling
Recommended Storage : Ambient temperatures