Learn More
Alfa Aesar™ Glassy Carbon Boats
Offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids
Manufacturer:  Alfa Aesar 40999KT
 View more versions of this product
Catalog No. AA40999KT
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
76mm x 33mm x 25mm
76mm
40mL vol.
25mm
Glassy Carbon Boat
33mm