Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Optical Windows

Available in different materials and dimensions

Manufacturer:  Alfa Aesar 45636KS

 View more versions of this product

Catalog No. AA45636KS


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Thickness: 2mm (0.08 in.)
50.8mm
Gallium arsenide
2 in.
Surface finish: 60/40
Optical Window
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit