Alfa Aesar™ Polyethylene Glove Bags
For handling air and moisture sensitive, high purity chemicals and materials
Manufacturer: Alfa Aesar 93727LK
 View more versions of this product
Catalog No. AA93727LK
  Check Availability
Add to Cart
    Specifications
    Includes 1/bag
    Size 94 x 69 x 46cm; 85cm entrance
    For Use With (Application) For handling air, moisture sensitive, high purity chemicals and materials
  • Description & Specifications
Related Products
Product Content Feedback