Learn More
Alfa Aesar™ Polyethylene Glove Bags
For handling air and moisture sensitive, high purity chemicals and materials
Manufacturer:  Alfa Aesar 92727LL
 View more versions of this product
Catalog No. AA92727LL
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
69 x 69 x 38cm; 57cm entrance
4/bag
For handling air, moisture sensitive, high purity chemicals and materials