Learn More
Alfa Aesar™ Quant™ Total Hardness Test Strips
Manufacturer:  Alfa Aesar 35377QA
 View more versions of this product
Catalog No. AA35377QA
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1 Kit
Total Hardness Test Strips
50 to 410mg/L (ppm) or 5 to 26°e