Learn More
Peroxide, Quantofix™ Test Strips, Thermo Scientific Chemicals
Supplier:  Thermo Scientific Chemicals 023284QA
Catalog No. AA23284QA
Order in Bulk
Request bulk or custom formats
Add to Cart
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Measures 1 to 25ppm peroxide in ether
Peroxide Test Strip
1 Kit
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
Recommended Storage : Ambient temperatures