Alfa Aesar™ Silica Gel 60, 70 to 150 Mesh

Manufacturer: Alfa Aesar 4272318

 View more versions of this product

Catalog No. AA4272318

  Check Availability
Add to Cart

    Specifications

    EINECS Number 231-545-4
    Product Type Silica gel
    Packaging Bottle
    pH 6.5 to 7.5
    Quantity 50g
    Surface Area (Metric) 470 to 570m2/g
    Material Silica
    Particle Size -70 to 200 mesh
    Pore Size 60Å

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit