Learn More
Thin Insulating Tubes, Thermo Scientific™
Oxide ceramics tubes resistant to corrosive liquids and gases
Supplier:  Thermo Scientific Chemicals 31489DW
 View more versions of this product
Catalog No. AA31489DW
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Inert to oxidation
  • Not subject to radiation damage
  • Wear resistant and hard
  • With both ends open
TRUSTED_SUSTAINABILITY
Specifications
Specifications
300 mm
1.5mm
2mm
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
TSCA : U
Recommended Storage : Ambient temperatures