Alfa Aesar™ TLC Grade 12 Micron APS Silica Gel

Manufacturer: Alfa Aesar 4276518

 View more versions of this product

Catalog No. AA4276518

  Check Availability
Add to Cart

    Specifications

    EINECS Number 231-545-4
    Product Type Silica gel
    Grade TLC
    Surface Area (Metric) 500 to 600m2/g
    Material Silica
    Pore Size 60Å
    pH 6.5 to 7.5
    Quantity 50g
    Particle Size 12μ

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit