Learn More
Alfa Aesar™ TLC Grade 12 Micron APS Silica Gel
Manufacturer:  Alfa Aesar 42765A7
 View more versions of this product
Catalog No. AA42765A7
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Silica
60Å
6.5 to 7.5
500 to 600m2/g
12μ
TLC
5kg
Silica gel