Learn More
Thermo Scientific Chemicals Wig-L-Bug™ Accessories
For use with Digital Wig-L-Bug Mixing/Grinding Mill
Supplier:  Thermo Scientific Chemicals 42628KJ
 View more versions of this product
Catalog No. AA42628KJ
Order in Bulk
Request bulk or custom formats
Add to Cart
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Polystyrene vial with polyethylene slip-on cap; 1 ft. long x 0.5 in. dia.
  • Polystyrene vial with polyethylene slip-on cap; 2 ft. long x 0.5 in. dia.
  • Adapter for 2 in. long, 0.5 in. diameter vials
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Accessory for Digital Wig-L-Bug mixing/grinding mill
1 ft.
5/Pk.
Polystyrene Vial
Product Suggestions
Product Suggestions