Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Wig-L-Bug™ Mixing/Grinding Mill Accessory, Ball Pestles, 9.52mm Dia.

For use with Wig-L-Bug Mixing/Grinding Mill (AA42627KT)

$27.55 - $70.70

Specifications

Product Type Ball Pestle
Diameter (Metric) Outer 9.52mm
For Use With (Equipment) Wig-L-Bug Mixing/Grinding Mill (AA42627KT)
View More Specs

Products 2
Catalog Number Mfr. No. Item Description Price Quantity & Availability  
Catalog Number Mfr. No. Item Description Price Quantity & Availability  
AA42631KM
View Documents
Alfa Aesar™
42631KM
Quantity: 25/Pk.
Pack for $27.55
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA42631KN
View Documents
Alfa Aesar™
42631KN
Quantity: 100/Pk.
Pack for $70.70
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

Specifications

Specifications

Ball Pestle
9.52mm
Wig-L-Bug Mixing/Grinding Mill (AA42627KT)
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit