Learn More
Alfa Aesar™ Yttria Stabilized Zirconium Oxide Conical Crucibles, High Form
Manufacturer:  Alfa Aesar 41290KF
 View more versions of this product
Catalog No. AA41290KF
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yttria Stabilized Zirconium Oxide Conical Crucibles, High Form
72mm
100mL
O.D. (Top): 62mm; O.D. (Base): 41mm
1 Piece